missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ Tris-Glycine Native Sample Buffer (2X)
Description
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Chemical Name or Material | Sample Loading Buffer |
| Concentration | 2X |
| For Use With (Equipment) | Mini Precast Protein Gels |
| pH | 8.6 |
| Physical Form | Liquid |
| Product Line | Novex |
| Quantity | 20 mL |
| Formulation | Glycerol 56-81-5 (20-30) |
For Research Use Only. Not for use in diagnostic procedures.