missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Novex™ Tricine XCell SureLock™ Mini-Cell Welcome Pack, 10-20%

Catalog No. EC6625E
Encompass
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
EC6625E 12-well
EC6625D 10-well
EC6625F 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EC6625E Supplier Thermo Scientific™ Supplier No. EC6625E
Only null left

The Novex Tricine Welcome Pack provides all of the necessary Novex Tricine gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.

The Novex Tricine Welcome Pack provides all of the necessary Novex Tricine gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.

The Novex Tricine Welcome Pack contains:
• XCell SureLock Mini-Cell (EI0001)
• Novex Tricine Mini Gels (2 boxes, 20 gels)
• Novex Tricine SDS Running Buffer, 10X (LC1675)
• Novex Tricine SDS Sample Buffer, 2X (LC1676)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• Spectra Multicolor Low Range Protein Ladder (26628)

About the XCell SureLock Mini-Cell 
The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About Novex Tricine gels
Novex Tricine gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow specifically for peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and de-stacking of low molecular weight proteins and higher resolution of smaller peptides.

Transfer
For transfer of proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

Specifications

Gel Percentage 10 to 20%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tricine
Separation Range 2.5 to 200 kDa
Wells 12-well
Quantity 1 Welcome Pk.
Shipping Condition Wet Ice
Product Line Novex
For Use With (Equipment) XCell SureLock Mini-Cell

For Research Use Only. Not for use in diagnostic procedures.