missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ TBE-Urea Sample Buffer (2X)

Catalog No. lc6876
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC6876 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC6876 Supplier Invitrogen™ Supplier No. LC6876

Compatible with agarose and polyacrylamide gels

For optimum performance, Novex™ TBE-Urea Sample Buffer is recommended for use with Novex™ TBE-urea gels.

  • Contains urea and density agent Ficoll™
  • Yields sharper, straighter bands than conventional density agents, plus tracking dyes bromophenol blue and xylene cyanol

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Wet Ice

Specifications

Buffer Sample Loading Buffer
Concentration 2 X
Content And Storage Store at 2°C to 8°C
Gel Compatibility Polyacrylamide Gels, Agarose Gels
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Label or Dye Bromophenol Blue, Xylene Cyanol
Product Line Novex
Product Type TBE Sample Buffer
Quantity 10 mL
Shipping Condition Wet Ice
My Novex TBE buffer has precipitated out of solution. What can I do?

If a slight turbidity develops, the fine precipitate can be dissolved by autoclaving for 5 minutes at 110°C. Do not autoclave in the container supplied. This treatment has no deleterious effect on the buffering properties of TBE.

Can a sample buffer with formamide be used with the Invitrogen TBE-Urea system?

There are many sample buffer formulations used, however we have found a distinct difference in the band appearance depending on the sample buffer composition. After evaluating urea, formamide, and various buffer systems, we found that the sharpest, flattest bands were obtained with a urea, Ficoll, and TBE buffer solution. Sample buffers made with formamide resulted in fuzzy, indistinct bands.

What can be used to stain Invitrogen precast TBE-Urea or TBE gels?

(1) Ethidium bromide: Soak gel in a 2 µg/mL solution of ethidium bromide in ultrapure water for 20 min. Destain by rinsing with three successive 10-min rinses of ultrapure water. Visualize bands under UV light. The gel must be removed from the cassette prior to visualization of the DNA under a UV light. Because polyacrylamide quenches the fluorescence of ethidium bromide, it is not possible to detect bands that contain less than about 10 ng of DNA by this method. SilverXpress and SYBR stains will provide greater detection sensitivity.

(2) Invitrogen SilverXpress Stain: Follow the standard procedure from the instruction booklet for staining TBE or TBE-Urea gels.

(3) SYBR Green I/II Nucleic Acid Gel Stain: See the SYBR Green Staining Manual for protocol details.

For Research Use Only. Not for use in diagnostic procedures.