missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuPAGE™ MES SDS Running Buffer (20X)
Made with high-purity reagents and are strictly quality controlled
$147.00 - $1106.00
Specifications
| Chemical Name or Material | Running Buffer |
|---|---|
| Concentration | 20X |
| pH | 7.3 |
| Physical Form | Liquid |
| Product Line | NuPAGE |
Includes: NuPAGE MES SDS Running Buffer (20X)
Description
NuPAGE MES SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE MES SDS Running Buffer is recommended for separating small- to medium-sized proteins.
- Use of MES buffer allows proteins to run faster
- Ensures high-quality, consistent electrophoresis results
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
| Running Buffer | |
| 7.3 | |
| NuPAGE |
| 20X | |
| Liquid | |
| 1X: 50 mM MES, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.3 |
For Research Use Only. Not for use in diagnostic procedures.