missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 2D-well DFS Item

Separation Range: 20 to 200 kd; Separation Type: Denaturing, Native; For Use With: Mini Gel Tank, XCell SureLock Mini

Supplier:  Invitrogen™ EC6026BOX

Catalog No. EC6026BOX


Only null left
Explore available promotions

Includes: One box of 10 gels

Description

Description

  • Provide reproducible separation of a wide range of proteins into well-resolved bands
  • Individually packaged gels for optimal experiment design
  • Compatible with most protein standards; usable for native and denaturing protein assays
  • For best results, most Novex Tris-Glycine gels should be stored at 4°C and used within eight weeks of purchase
  • Run proteins in native or denatured form
  • Do not contain SDS and can be used to run proteins in native or in denatured form

2D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

One box contains 10 gels. Store in refrigerator (2–8°C). Shelf life up to 12 months, depending on gel type.
Mini
Tris-Glycine
20 to 200 kDa
2D-well
10 gels (1 box)
1.0 mm
8 cm
8 cm
Novex
Mini Gel Tank, XCell SureLock Mini-Cell
4 to 20%
1.0 mm
Protein
Denaturing, Native
Molecular Weight
Wet Ice
2D Well
8 cm
8 cm
Gradient Page
ZOOM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.