Learn More
Invitrogen™ 20X Bolt™ MOPS SDS Running Buffer
Description
About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.
Bolt™ gel run times may vary depending upon the type of running buffer used.
Order Info
Shelf-life is 6 months, when stored properly.
Specifications
Specifications
| Chemical Name or Material | Running Buffer |
| Concentration | 20X |
| For Use With (Equipment) | Bolt |
| Physical Form | Liquid |
| Product Line | Bolt |
| Quantity | 5 L |
For Research Use Only. Not for use in diagnostic procedures.