missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ X-Gal Solution

Catalog No. 710773
Change view
Click to view available options
Quantity:
3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
710773 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 710773 Supplier MilliporeSigma™ Supplier No. 710773

Convenient for blue/white screening

  • The β-galactosidase substrate X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside) is a chromogenic stain for β-gal activity, and is commonly used to distinguish between recombinants and nonrecombinants by lacZ α-complementation with appropriate vectors and hosts
  • X-Gal Solution is provided as a convenient 40mg/mL concentrate in DMSO, ready for dilution into culture medium or appropriate buffers

Specifications

Content And Storage Store at −20°C
Quantity 3 mL