missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ T4 DNA Polymerase, LIC-Qualified

Catalog No. 700993
Change view
Click to view available options
Quantity:
250 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
700993 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 700993 Supplier MilliporeSigma™ Supplier No. 700993

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For any ligation or cloning application

  • Has been qualified for ligation-independent cloning (LIC) using unique assay to verify its performance

Specifications

Content And Storage -20°C
Quantity 250 U
Product Type T4 DNA Polymerase, LIC-Qualified

For Research use Only