missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ SpinPrep™ PCR Clean-up Kit

Catalog No. 709763
Change view
Click to view available options
SpinPrep™ PCR Clean-up Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
709763 SpinPrep™ PCR Clean-up Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 709763 Supplier MilliporeSigma™ Supplier No. 709763

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for rapid purification of DNA amplified in PCR.

EMD Millipore Novagen™ SpinPrep™ PCR Clean-up Kit removes DNA polymerases, dNTPs, salts gt; 99% of primers.

  • Addition of binding buffer adsorps DNA to silica membrane in spin column format
  • DNA is eluted in low-salt buffer
  • Removes DNA polymerases, dNTPs, salts, > 99% of primers so that they do not Interfere with downstream applications such as cloning, sequencing, or labeling.
  • PCR products from 100 bp to > 12,000 bp can be cleaned
  • Recoveries of amplified DNA in the range of 60-90% under standard conditions
  • Packaging: Glass Bottle

Specifications

Content And Storage 2°C to 8°C
Format Kit
Isolation Technology Silica Membrane Technology
No. of Reactions 100 Reactions/Kit
Product Type PCR Clean-up Kit