Learn More
MilliporeSigma™ Novagen™ S-protein Agarose
Description
EMD Millipore Novagen™ S-protein Agarose binds in both column or batch mode.
- Elution is done under slightly denaturing conditions or using specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins
- Capacity varies is based on size folding characteristics of given target protein
- Packaging: Glass Bottle
Specifications
Specifications
| Quantity | 2 mL |