missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ S-protein Agarose

Catalog No. 697043
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
697043 2 mL
697044 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 697043 Supplier MilliporeSigma™ Supplier No. 697043

Retains specifically S·Tag™ fusion proteins.

EMD Millipore Novagen™ S-protein Agarose binds in both column or batch mode.

  • Elution is done under slightly denaturing conditions or using specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins
  • Capacity varies is based on size folding characteristics of given target protein
  • Packaging: Glass Bottle

Specifications

Quantity 2 mL