missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ rLysozyme™ Solution
For lysis of Gram-negative bacteria
$186.00
Specifications
| Concentration | 27 to 33K U/μL |
|---|---|
| Components | Supplied as a ready-to-use solution at a concentration range of 27 to 33KU/μL in 50% glycerol containing 50mM Tris-HCl, 0.1M NaCl, 0.1mM EDTA, 1 mM DTT, 0.1% TRITON™ X-100 |
| pH | 7.5 |
| Content And Storage | -20°C |
| Product Type | Stabilized recombinant lysozyme |
Description
- Contains highly purified and stabilized recombinant lysozyme
- Catalyzes hydrolysis of N-acetylmuramide linkages in bacterial cell walls
- Specific activity of rLysozyme (1700KU/mg) for E. coli lysis is 250 times greater than that of chicken egg white lysozyme
- Optimally active at physiological pH
Specifications
| 27 to 33K U/μL | |
| 7.5 | |
| Stabilized recombinant lysozyme |
| Supplied as a ready-to-use solution at a concentration range of 27 to 33KU/μL in 50% glycerol containing 50mM Tris-HCl, 0.1M NaCl, 0.1mM EDTA, 1 mM DTT, 0.1% TRITON™ X-100 | |
| -20°C |
For Research Use Only