missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ PopCulture™ Reagent

Catalog No. p-7078303
Change view
Click to view available options
Quantity:
15 mL
75 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
710924 75 mL
710923 15 mL
710925 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 710924 Supplier MilliporeSigma™ Supplier No. 710924
Only null left

Formulated to extract proteins from E. coli cells directly in culture medium.

EMD Millipore Novagen™ PopCulture™ Reagent is buffered mixture of concentrated detergents.

  • Perforates E. coli cell wall without denaturing soluble protein protects protein from pH extremes
  • Assayed directly or purified by adding affinity matrix
  • Performed in single tube without need for columns or centrifugation
  • Applications:
    • High throughput (HT) robotic processing of samples for proteomics research
    • Packaging: Glass Bottle

Specifications

Content And Storage 15°C to 30°C
Quantity 75 mL