missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ pET-32 Ek/LIC Vector Kit

Catalog No. 690763
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
690763 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 690763 Supplier MilliporeSigma™ Supplier No. 690763

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for cloning high-level expression of target proteins.

EMD Millipore Novagen™ pET-32 Ek/LIC Vector Kit is Ek/LIC version of pET-32b(+).

  • Target proteins fused with 109aa Trx Tag™ thioredoxin protein, His Tag™ S™Tag™ coding sequences that are cleavable with enterokinase (Ek) protease
  • Optional C-terminal His Tag coding sequence is compatible with purification, detection, quantification
  • Packed in Glass Bottle

Specifications

Type pET-32 Ek/LIC Vector Kit
Sufficient For 20 Reactions
Quantity 1 Kit
Content And Storage ≤-70°C