missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Pellet Paint™ Co-Precipitant

Catalog No. 690494
Change view
Click to view available options
No. of Reactions:
1000
125
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
690494 1000
690493 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 690494 Supplier MilliporeSigma™ Supplier No. 690494

Compatible with most molecular biology procedures and is free of contaminating nucleic acids and nucleolytic enzymes

Direct visualization and tracking of precipitated material

Specifications

Content And Storage Store at −20°C
Label or Dye Fluorescent dye-labeled
No. of Reactions 1000
Form Liquid