Learn More
MilliporeSigma™ Novagen™ Pellet Paint™ Co-Precipitant
Compatible with most molecular biology procedures and is free of contaminating nucleic acids and nucleolytic enzymes
Supplier:  MilliporeSigma™ 690493
 View more versions of this product
Catalog No. 690493
Only null left
Description
Description
Compatible with most molecular biology procedures and is free of contaminating nucleic acids and nucleolytic enzymes
Direct visualization and tracking of precipitated material
Specifications
Chemical Identifiers
Store at −20°C
Fluorescent dye-labeled
Liquid
Specifications
Store at −20°C
Fluorescent dye-labeled
125
Liquid
Product Suggestions
Product Suggestions