missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ Overnight Express™ Autoinduction System 2
Description
Includes
- 100mL OnEx™ Solution 1
- 125mL OnEx Solution 2
- 1mL OnEx Solution 3
- 1mL OnEx Solution 4
- 100mL OnEx Solution 5
- 100mL OnEx Solution 6
EMD Millipore Novagen™ Overnight Express™ Autoinduction System 2 provides complete, chemically defined medium for high-level protein expression with pET System.
- High cell densities and protein expression levels
- Chemically defined medium, minimal variability
- No need for monitor cell growth rate or add inducer
- Used for Se-Met labeling of proteins to be crystallized for x-ray diffraction studies
- Induction of numerous expression clones simultaneously
- Compatible with cultures grown in flasks, culture tubes, and deep-well plates
- Minimal sample handling
- Packaging: Glass Bottle
Specifications
Specifications
| Quantity | 5 L Kit |
| Content And Storage | Multiple storage requirements |