Learn More
MilliporeSigma™ Novagen™ NovaTaq™ Hot Start Master Mix Kits
Premixed 2X hot start PCR components for convenience and reproducibility
$370.00 - $1,660.00
Specifications
Content And Storage Ready-to-use 2X mixture of NovaTaq Hot Start DNA Polymerase, ultrapure deoxynucleotides, and reaction buffer with MgCl2, PCR Grade Water, MgCl2 for optimizing Mg2+ concentrations
Product Type Hot start master mix kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
716764
View Documents Promotion Details
MilliporeSigma™
716764
1000 rxn
Each for $1,660.00
Only null left
716763
View Documents Promotion Details
MilliporeSigma™
716763
200 rxn
Each for $370.00
Only null left
Description
Description
  • Simplifies set-up for PCR and offers time savings, consistency, and minimal risk of contamination
  • Provide sufficient components for 200 or 1000 standard 50μL amplification reactions
Specifications
Specifications
Ready-to-use 2X mixture of NovaTaq Hot Start DNA Polymerase, ultrapure deoxynucleotides, and reaction buffer with MgCl2, PCR Grade Water, MgCl2 for optimizing Mg2+ concentrations
Hot start master mix kit