missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ Ni-NTA HisBind™ Superflow™ Resin
Description
EMD Millipore Novagen™ Ni-NTA HisBind™ Superflow™ Resin is medium-pressure compatible version of Ni-NTA HisBind.
- Medium-pressure compatible version of Ni-NTA HisBind
- Binding capacity: 5 to 50mg HisTag™ fusion protein per mL resin
- Packaging: Glass Bottle
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Quantity | 100 mL |