missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Insect PopCulture™ Reagent

Catalog No. 711873
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
711873 50 mL
711874 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 711873 Supplier MilliporeSigma™ Supplier No. 711873

Extracts protein directly from insect cell cultures.

EMD Millipore Novagen™ Insect PopCulture™ Reagent is detergent-based lysis reagent without the need for centrifugation.

  • Recovers both protein released into medium and intracellular protein
  • Increases processing efficiency and target protein yields
  • Amenable for automated expression-level screening
  • Compatible with Ni-NTA His·Bind™ affinity purification method
  • Packaging: Glass Bottle

Specifications

Content And Storage 2°C to -8°C
Quantity 50 mL