missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ His Bind™ Resin

Catalog No. 696703
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
696703 10 mL
696704 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 696703 Supplier MilliporeSigma™ Supplier No. 696703

Suited to small to medium scale purifications.

EMD Millipore Novagen™ His Bind™ Resin is an uncharged IDA agarose.

  • Binding capacity is approximately 8mg/mL bed volume
  • Uses either gravity flow columns or batch methods

Specifications

Content And Storage 2°C to 8°C
Quantity 10 mL
Type Chromatography Media