missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ GST Bind™ Resin

Catalog No. 705415
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
705415 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 705415 Supplier MilliporeSigma™ Supplier No. 705415

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Utilizes 14-atom spacer arm to covalently attach reduced glutathione via sulfide linkage.

EMD Millipore Novagen™ GST·Bind™ Resin has high degree of substitution of glutathione.

  • Ensures high binding capacity
  • Yield: 5 to 8mg/mL settled resin
  • Can be re-used up to 6 times without loss of capacity

Specifications

Content And Storage 2°C to 8°C. Do not freeze.
Quantity 25 mL