Learn More
MilliporeSigma™ Novagen™ Ekapture™ Agarose
Intended for rapid affinity-based capture.
$876.00 - $876.00
Description
EMD Millipore Novagen™ Ekapture™ Agarose is used for removal of enterokinase.
- Removed with >99% efficiency from reaction by affinity capture
Specifications
| 2°C to 8°C | |
| Agarose |