missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Chicken Egg White Lysozyme Solution

Catalog No. 714123
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
714123 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 714123 Supplier MilliporeSigma™ Supplier No. 714123

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For purification of both DNA, protein, and Gram-positive bacterial lysis

Specifications

Concentration 10 mg/mL
Components Supplied as 10mg/mL lysozyme in 25mM sodium acetate with 50% glycerol
pH 4.5
Content And Storage -20°C
Quantity 10 mL
Product Type Chicken Egg White Lysozyme Solution

For Research use Only