missing translation for 'onlineSavingsMsg'
Learn More

Noscapine Hydrochloride Hydrate 98.0+%, TCI America™

Supplier:  TCI America N09635G

 View more versions of this product

Catalog No. N09635G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Noscapine Hydrochloride Hydrate
200°C
C22H24ClNO7
5 g
MFLVZFXCSKVCSH-OQICONIUSA-N
(5S)-5-(4,5-dimethoxy-3-oxo-1,3-dihydro-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride
20845981
≥98.0% (HPLC,T)
912-60-7
White
MFCD00082546
narcotine hydrochloride, noscapine hydrate hydrochloride, narcotine hydrochloride hydrate, c22h23no7.clh.h2o, noscapine hydrochloride monohydrate, noscapine hydrochloride hydrate jp17, noscapine hydrochloride hydrate, analytical standard
[Cl-].COC1=CC=C2C(OC(=O)C2=C1OC)[C@H]1[NH+](C)CCC2=CC3=C(OCO3)C(OC)=C12
449.88
449.88
Crystalline Powder

Chemical Identifiers

912-60-7
449.88
MFLVZFXCSKVCSH-OQICONIUSA-N
20845981
[Cl-].COC1=CC=C2C(OC(=O)C2=C1OC)[C@H]1[NH+](C)CCC2=CC3=C(OCO3)C(OC)=C12
C22H24ClNO7
MFCD00082546
narcotine hydrochloride, noscapine hydrate hydrochloride, narcotine hydrochloride hydrate, c22h23no7.clh.h2o, noscapine hydrochloride monohydrate, noscapine hydrochloride hydrate jp17, noscapine hydrochloride hydrate, analytical standard
(5S)-5-(4,5-dimethoxy-3-oxo-1,3-dihydro-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : NP7225000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions