CAS RN 912-60-7
CAS RN 912-60-7
IUPAC Name:
(5S)-5-(4,5-dimethoxy-3-oxo-1,3-dihydro-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride
Synonyms:
(5S)-5-(4,5-dimethoxy-3-oxo-1H-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride
Molecular Weight (g/mol):
449.88
Molecular Formula:
C22H24ClNO7
InChi Key:
MFLVZFXCSKVCSH-OQICONIUSA-N
SMILES:
[Cl-].COC1=CC=C2C(OC(=O)C2=C1OC)[C@H]1[NH+](C)CCC2=CC3=C(OCO3)C(OC)=C12
1
–
2
of
2
results
2
Noscapine Hydrochloride Hydrate 98.0+%, TCI America™
CAS: 912-60-7 Molecular Formula: C22H24ClNO7 Molecular Weight (g/mol): 449.88 MDL Number: MFCD00082546 InChI Key: MFLVZFXCSKVCSH-OQICONIUSA-N Synonym: narcotine hydrochloride,noscapine hydrate hydrochloride,narcotine hydrochloride hydrate,c22h23no7.clh.h2o,noscapine hydrochloride monohydrate,noscapine hydrochloride hydrate jp17,noscapine hydrochloride hydrate, analytical standard PubChem CID: 20845981 IUPAC Name: (5S)-5-(4,5-dimethoxy-3-oxo-1,3-dihydro-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride SMILES: [Cl-].COC1=CC=C2C(OC(=O)C2=C1OC)[C@H]1[NH+](C)CCC2=CC3=C(OCO3)C(OC)=C12