missing translation for 'onlineSavingsMsg'
Learn More

Noscapine Hydrochloride Hydrate 98.0+%, TCI America™

Catalog No. p-7134052
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09635G Supplier TCI America Supplier No. N09635G

Chemical Identifiers

CAS 912-60-7
Molecular Formula C22H24ClNO7
Molecular Weight (g/mol) 449.88
MDL Number MFCD00082546
InChI Key MFLVZFXCSKVCSH-OQICONIUSA-N
Synonym narcotine hydrochloride, noscapine hydrate hydrochloride, narcotine hydrochloride hydrate, c22h23no7.clh.h2o, noscapine hydrochloride monohydrate, noscapine hydrochloride hydrate jp17, noscapine hydrochloride hydrate, analytical standard
PubChem CID 20845981
IUPAC Name (5S)-5-(4,5-dimethoxy-3-oxo-1,3-dihydro-2-benzofuran-1-yl)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium chloride
SMILES [Cl-].COC1=CC=C2C(OC(=O)C2=C1OC)[C@H]1[NH+](C)CCC2=CC3=C(OCO3)C(OC)=C12

Specifications

Melting Point 200°C
Color White
Quantity 5 g
Formula Weight 449.88
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Noscapine Hydrochloride Hydrate
RTECSNumber NP7225000
TSCA No
Recommended Storage Refrigerator