Learn More
Norethisterone, Thermo Scientific Chemicals
$187.51 - $513.82
Chemical Identifiers
| CAS | 68-22-4 |
|---|---|
| Molecular Formula | C20H26O2 |
| Molecular Weight (g/mol) | 298.42 |
| InChI Key | VIKNJXKGJWUCNN-XGXHKTLJSA-N |
| Synonym | norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone |
| PubChem CID | 6230 |
| ChEBI | CHEBI:7627 |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
| SMILES | CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC262890010
|
Thermo Scientific Chemicals
262890010 |
1 g |
Each for $187.51
|
|
|||||
|
AC262890050
|
Thermo Scientific Chemicals
262890050 |
5 g |
Each for $513.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 68-22-4 | |
| 298.42 | |
| norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone | |
| CHEBI:7627 | |
| CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34 |
| C20H26O2 | |
| VIKNJXKGJWUCNN-XGXHKTLJSA-N | |
| 6230 | |
| (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
Specifications
| 68-22-4 | |
| 0.5% max. (1 g, 105°C, 3 h) | |
| C20H26O2 | |
| norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone | |
| VIKNJXKGJWUCNN-XGXHKTLJSA-N | |
| (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one | |
| 6230 | |
| 298.42 | |
| Norethisterone |
| 205.0°C to 206.0°C | |
| Authentic | |
| 1 g | |
| Solubility in water: insoluble. Other solubilities: soluble in chloroform, dioxane | |
| CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34 | |
| 298.42 | |
| CHEBI:7627 | |
| ≥97.5% (HPLC) |
Safety and Handling
GHS H Statement Suspected of causing cancer. May damage fertility. May damage the unborn child. May cause harm to breast-fed children.
GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention. Avoid contact during pregnancy/while nursing.
GHS Signal Word: Danger
EINECSNumber : 200-681-6
RUO – Research Use Only