accessibility menu, dialog, popup

UserName

H3C(S)(S)(R)O(R)(S)(R)HOHC

CAS RN 68-22-4

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-1-ethynyl-1-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: norethindrone noretisterona [inn-spanish] norethindrone, (1 β)-isomer noresthisterone norcept norethindrone (substance) loestrin 24 fe (1 β)-norethindrone ethinylnortestosterone jenest
Molecular Weight (g/mol): 298.43
Molecular Formula: C20H26O2
InChi Key: VIKNJXKGJWUCNN-XGXHKTLJSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)