missing translation for 'onlineSavingsMsg'
Learn More

Nordihydroguaiaretic Acid 97.0+%, TCI America™

Catalog No. p-7126438
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08001G 1 g
D08005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08001G Supplier TCI America Supplier No. D08001G
Only null left

Chemical Identifiers

CAS 500-38-9
Molecular Formula C18H22O4
Molecular Weight (g/mol) 302.37
MDL Number MFCD00002206
InChI Key HCZKYJDFEPMADG-UHFFFAOYSA-N
Synonym nordihydroguaiaretic acid, ndga, dihydronorguaiaretic acid, norhydroguaiaretic acid, nordihydroguairaretic acid, norguaiaretic acid, dihydro, 1,2-benzenediol, 4,4'-2,3-dimethyl-1,4-butanediyl bis, dinorguaiaretic acid, dihydro, 4,4'-2,3-dimethyltetramethylene dipyrocatechol, 2,3-bis 3,4-dihydroxyphenylmethyl butane
PubChem CID 4534
ChEBI CHEBI:7625
IUPAC Name 4-[4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
SMILES CC(CC1=CC(=C(C=C1)O)O)C(C)CC2=CC(=C(C=C2)O)O

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 1 g
Formula Weight 302.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nordihydroguaiaretic Acid
RTECSNumber UX1750000
TSCA No