accessibility menu, dialog, popup

UserName

CAS RN 500-38-9

IUPAC Name: 4-[(2R,3S)-4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
Synonyms: 4-[(2R,3S)-4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol masoprocolum NDGA dinorguaiaretic acid, dihydro- masoprocol (substance) masoprocol (product) dihydronorguaiaretic acid masoprocol preparation norguaiaretic acid, dihydro- actinex
Molecular Weight (g/mol): 302.37
Molecular Formula: C18H22O4
InChi Key: HCZKYJDFEPMADG-TXEJJXNPSA-N
SMILES: C[C@@H](CC1=CC=C(O)C(O)=C1)[C@H](C)CC1=CC=C(O)C(O)=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)