missing translation for 'onlineSavingsMsg'
Learn More

Nordihydroguaiaretic acid, 97%

Catalog No. AAL0314903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0314903 1 g
AAL0314906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0314903 Supplier Thermo Scientific Chemicals Supplier No. L0314903

CAS: 500-38-9 | C18H22O4 | 302.37 g/mol

Nordihydroguaiaretic acid is used in the treatment of multiple diseases, including cardiovascular diseases, neurological disorders and cancers. It is also used as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 100% ethanol, methanol, acetone, dimethylsulfoxide, dimethylformamide, ethanol (5% w/v), glycol, PEG, and slightly soluble in hot water. Insoluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 500-38-9
Molecular Formula C18H22O4
Molecular Weight (g/mol) 302.37
MDL Number MFCD00002206
InChI Key HCZKYJDFEPMADG-UHFFFAOYSA-N
Synonym nordihydroguaiaretic acid, ndga, dihydronorguaiaretic acid, norhydroguaiaretic acid, nordihydroguairaretic acid, norguaiaretic acid, dihydro, 1,2-benzenediol, 4,4'-2,3-dimethyl-1,4-butanediyl bis, dinorguaiaretic acid, dihydro, 4,4'-2,3-dimethyltetramethylene dipyrocatechol, 2,3-bis 3,4-dihydroxyphenylmethyl butane
PubChem CID 4534
ChEBI CHEBI:7625
IUPAC Name 4-[4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
SMILES CC(CC1=CC(=C(C=C1)O)O)C(C)CC2=CC(=C(C=C2)O)O

Specifications

Melting Point 184°C to 188°C
Odor Odorless
Quantity 1 g
Beilstein 2056825
Merck Index 14,6693
Solubility Information Soluble in 100% ethanol,methanol,acetone,dimethylsulfoxide,dimethylformamide,ethanol (5% w/v),glycol,PEG,and slightly soluble in hot water. Insoluble in water.
Formula Weight 302.37
Percent Purity 97%
Chemical Name or Material Nordihydroguaiaretic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-903-0
RTECSNumber UX1750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only