missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonacosane 98.0+%, TCI America™
$121.16 - $562.95
Chemical Identifiers
| CAS | 630-03-5 |
|---|---|
| Molecular Formula | C29H60 |
| Molecular Weight (g/mol) | 408.80 |
| MDL Number | MFCD00009356 |
| InChI Key | IGGUPRCHHJZPBS-UHFFFAOYSA-N |
| PubChem CID | 12409 |
| ChEBI | CHEBI:7613 |
| IUPAC Name | nonacosane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N0167100MG
|
TCI America
N0167100MG |
100 mg |
Each for $121.16
|
|
|||||
|
N01671G
|
TCI America
N01671G |
1 g |
Each for $562.95
|
|
|||||
Chemical Identifiers
| 630-03-5 | |
| 408.80 | |
| IGGUPRCHHJZPBS-UHFFFAOYSA-N | |
| CHEBI:7613 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
| C29H60 | |
| MFCD00009356 | |
| 12409 | |
| nonacosane |
Specifications
| 630-03-5 | |
| C29H60 | |
| White | |
| IGGUPRCHHJZPBS-UHFFFAOYSA-N | |
| nonacosane | |
| 100 mg | |
| CHEBI:7613 | |
| ≥98.0% (GC) |
| Powder/Crystals | |
| MFCD00009356 | |
| 64°C | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 408.80 | |
| 12409 | |
| 408.80 | |
| Nonacosane |
Safety and Handling
EINECSNumber : (2)-0010
TSCA : Yes