missing translation for 'onlineSavingsMsg'
Learn More

Nonacosane 98.0+%, TCI America™

Supplier:  TCI America N0167100MG

 View more versions of this product

Catalog No. N0167100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nonacosane
Powder/Crystals
MFCD00009356
64°C
CCCCCCCCCCCCCCCCCCCCCCCCCCCCC
408.80
12409
408.80
630-03-5
C29H60
White
IGGUPRCHHJZPBS-UHFFFAOYSA-N
nonacosane
100 mg
CHEBI:7613
≥98.0% (GC)

Chemical Identifiers

630-03-5
408.80
IGGUPRCHHJZPBS-UHFFFAOYSA-N
CHEBI:7613
CCCCCCCCCCCCCCCCCCCCCCCCCCCCC
C29H60
MFCD00009356
12409
nonacosane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0010

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions