Learn More
Nitroterephthalic acid, 99%
CAS: 610-29-7 | C8H5NO6 | 211.129 g/mol
$92.32 - $1182.75
Chemical Identifiers
| CAS | 610-29-7 |
|---|---|
| Molecular Formula | C8H5NO6 |
| Molecular Weight (g/mol) | 211.129 |
| MDL Number | MFCD00007141 |
| InChI Key | QUMITRDILMWWBC-UHFFFAOYSA-N |
| Synonym | nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u |
| PubChem CID | 69122 |
| SMILES | C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2504306
|
Thermo Scientific Chemicals
B2504306 |
5 g |
Each for $92.32
|
|
|||||
|
AAB2504314
|
Thermo Scientific Chemicals
B2504314 |
25 g |
Each for $300.10
|
|
|||||
|
AAB2504322
|
Thermo Scientific Chemicals
B2504322 |
100 g |
Each for $1,182.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 610-29-7 | |
| 211.129 | |
| QUMITRDILMWWBC-UHFFFAOYSA-N | |
| 69122 |
| C8H5NO6 | |
| MFCD00007141 | |
| nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u | |
| C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O |
Specifications
| 610-29-7 | |
| C8H5NO6 | |
| 5 g | |
| QUMITRDILMWWBC-UHFFFAOYSA-N | |
| 211.129 | |
| 211.13 | |
| Nitroterephthalic acid |
| 270°C to 272°C | |
| MFCD00007141 | |
| nitroterephthalic acid, 2-nitroterephthalic acid, 2-nitrobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-nitro, terephthalic acid, nitro, 2-nitroterephthalicacid, 2-nitroterephthalate, 5-nitroterephthalic acid, acmc-1au2u | |
| C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O | |
| 69122 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-217-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only