missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Standard, 1000 ppm NO3- (226 ppm N), Ricca Chemical

Catalog No. p-3252009
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
54584 120 mL Natural Poly Bottle
545816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54584 Supplier Ricca Chemical Company Supplier No. 54584
Only null left

Chemical Identifiers

CAS 7757-79-1 , 7732-18-5 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1000ppm Nitrate,226ppm N
CAS Min % 98.0, 0.16
CAS Max % 100.0, 0.17
Equivalence 1 mL = 1 mg NO3-
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Nitrogen Standard