missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Standard, 1000 ppm NO3- (226 ppm N), Ricca Chemical

Supplier:  Ricca Chemical Company 545816

 View more versions of this product

Catalog No. 5458-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nitrogen Standard
7757-79-1,7732-18-5,7732-18-5
1000ppm Nitrate,226ppm N
1 mL = 1 mg NO3-
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
500 mL
CHEBI:63043
Natural Poly Bottle
98.0,0.16
100.0,0.17
KNO3
100°C
[K+].[O-][N+]([O-])=O
101.10
24434

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[K+].[O-][N+]([O-])=O
KNO3
MFCD00011409
24434
potassium nitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions