missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrate Nitrogen Standard, 100 ppm N (442.7 ppm NO3-), Ricca Chemical
$101.31 - $189.44
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.10 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium nitrate |
| SMILES | [K+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
5457-4
|
Ricca Chemical Company
54574 |
120 mL | Natural Poly Bottle |
Each for $101.31
|
|
||||
|
5457-16
|
Ricca Chemical Company
545716 |
500 mL | Natural Poly Bottle |
Each for $141.72
|
|
||||
|
R54570001A
|
Ricca Chemical Company
R54570001A |
1 L | Natural Poly Bottle |
Each for $189.44
|
|
||||
Chemical Identifiers
| 7757-79-1 | |
| 101.10 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| CHEBI:63043 | |
| [K+].[O-][N+]([O-])=O |
| KNO3 | |
| MFCD00011409 | |
| 24434 | |
| potassium nitrate |
Specifications
| Natural Poly Bottle | |
| 98.0,0.07 | |
| KNO3 | |
| 100°C | |
| [K+].[O-][N+]([O-])=O | |
| 101.10 | |
| 24434 | |
| Nitrogen Standard, 1 mL = 1 mg N as NO3 |
| 7757-79-1,7732-18-5 | |
| 100.0,0.07 | |
| MFCD00011409 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium nitrate | |
| 120 mL | |
| CHEBI:63043 |