missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Nitrogen Standard, 100 ppm N (442.7 ppm NO3-), Ricca Chemical

Supplier:  Ricca Chemical Company 545716

 View more versions of this product

Catalog No. 5457-16

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nitrogen Standard
7757-79-1,7732-18-5,7732-18-5
100ppm N as Nitrate (442.7ppm NO3)
1 mL = 0.1 mg N
KNO3
Colorless
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
500 mL
CHEBI:63043
Natural Poly Bottle
98.0,0.07
100.0,0.07
Liquid
MFCD00011409
100°C
[K+].[O-][N+]([O-])=O
101.10
24434

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[K+].[O-][N+]([O-])=O
KNO3
MFCD00011409
24434
potassium nitrate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions