missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Nitrogen Standard, 10 ppm N (44.27 ppm NO3-), Ricca Chemical

$127.91 - $191.22

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5456-4
View Documents
Ricca Chemical Company
54564
120 mL Natural Poly Bottle
Each for $127.91
Only null left
 
5456-16
View Documents
Ricca Chemical Company
545616
500 mL Natural Poly Bottle
Each for $191.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[K+].[O-][N+]([O-])=O
KNO3
MFCD00011409
24434
potassium nitrate

Specifications

Natural Poly Bottle
98.0,0.01
100.0,0.01
Liquid
MFCD00011409
100°C
[K+].[O-][N+]([O-])=O
101.10
24434
Nitrogen Standard
7757-79-1,7732-18-5,7732-18-5
10ppm N as Nitrate (44.27ppm NO3)
1 mL = 0.01 mg N
KNO3
Colorless
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
120 mL
CHEBI:63043
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'