missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrate Nitrogen Standard, 10 ppm N (44.27 ppm NO3-), Ricca Chemical
Supplier: Ricca Chemical Company 545616
Specifications
| Nitrogen Standard | |
| 7757-79-1,7732-18-5,7732-18-5 | |
| 10ppm N as Nitrate (44.27ppm NO3) | |
| 1 mL = 0.01 mg N | |
| KNO3 | |
| Colorless | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium nitrate | |
| 500 mL | |
| CHEBI:63043 |
| Natural Poly Bottle | |
| 98.0,0.01 | |
| 100.0,0.01 | |
| Liquid | |
| MFCD00011409 | |
| 100°C | |
| [K+].[O-][N+]([O-])=O | |
| 101.10 | |
| 24434 |
Chemical Identifiers
| 7757-79-1 | |
| 101.10 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| CHEBI:63043 | |
| [K+].[O-][N+]([O-])=O |
| KNO3 | |
| MFCD00011409 | |
| 24434 | |
| potassium nitrate |