missing translation for 'onlineSavingsMsg'
Learn More

Nitrocyclohexane 95.0+%, TCI America™

Catalog No. N01875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01875G 5 g
N018725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01875G Supplier TCI America Supplier No. N01875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1122-60-7
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00003843
InChI Key NJNQUTDUIPVROZ-UHFFFAOYSA-N
PubChem CID 14285
ChEBI CHEBI:74164
IUPAC Name nitrocyclohexane
SMILES C1CCC(CC1)[N+](=O)[O-]

Specifications

Melting Point -34°C
Color Yellow
Boiling Point 113°C
Quantity 5 g
UN Number 2810
Formula Weight 129.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Nitrocyclohexane
RTECSNumber GV6600000
TSCA Yes