accessibility menu, dialog, popup

UserName

CAS RN 1122-60-7

IUPAC Name: nitrocyclohexane
Synonyms: nitrocyclohexane
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: NJNQUTDUIPVROZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results