missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrite (as Nitrogen) Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N, 1mL = 3.28mg NO2- ), LabChem™
$62.46 - $143.14
Chemical Identifiers
| CAS | 7758-09-0 |
|---|---|
| Molecular Formula | KNO2 |
| Molecular Weight (g/mol) | 85.10 |
| MDL Number | MFCD00011408 |
| InChI Key | BXNHTSHTPBPRFX-UHFFFAOYSA-M |
| PubChem CID | 516910 |
| IUPAC Name | potassium nitrite |
| SMILES | [K+].[O-]N=O |
Chemical Identifiers
| 7758-09-0 | |
| 85.10 | |
| BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
| potassium nitrite |
| KNO2 | |
| MFCD00011408 | |
| 516910 | |
| [K+].[O-]N=O |
Specifications
| Poly Bottle | |
| 99.39,0.61 | |
| 1g/mL | |
| Liquid | |
| 1000 ppm ±10 ppm | |
| MFCD00011408 | |
| Soluble in water | |
| [K+].[O-]N=O | |
| 85.10 | |
| 516910 | |
| Certified | |
| as N |
| 7758-09-0,7732-18-5 | |
| Passes Test | |
| 1g/mL | |
| KNO2 | |
| KNO2 | |
| Colorless | |
| BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
| potassium nitrite | |
| 125 mL | |
| 85.1 | |
| Nitrite (as Nitrogen) Standard, 1000 ppm, (1mL = 1mg N, 1mL = 3.28mg NO2-) |
Safety and Handling
GHS H Statement
Harmful to aquatic life.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage : Refrigerate