missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, Ultratrace, 67-70%, Spectrum™ Chemical

Catalog No. p-7165958
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
Packaging:
PTFE Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612013 500 mL PTFE Bottle
18612011 1 L PTFE Bottle
18612012 2 L PTFE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612013 Supplier Spectrum Chemical Mfg Cor Supplier No. N1097500MLTF
Only null left

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

CAS Max % 1
Packaging PTFE Bottle
Quantity 500 mL
Percent Purity 67 to 70%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.