missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, Ultratrace, 67-70%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor N10971LTTF
Specifications
| 7697-37-2,7732-18-5 | |
| PTFE Bottle | |
| HNO3 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 67 to 70% |
| 1 | |
| 1 L | |
| MFCD00011349 | |
| O[N+]([O-])=O | |
| 63.01 | |
| High Purity Solvent |
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| O[N+]([O-])=O |
| HNO3 | |
| MFCD00011349 | |
| nitric acid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.