missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, Technical 42 Degrees Baume’, Reagents

Catalog No. 01336826
Change view
Click to view available options
Quantity:
2.2 L
600 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336826 2.2 L
01336825 600 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336826 Supplier Labchem Inc Supplier No. C51017002.2A
Only null left

Nitric Acid, Technical 42 Degrees Baume’, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate, Aqua fortis
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 0
Color Colorless to Slight Yellow
Boiling Point 121°C
Packaging Poly Bottle
Specific Gravity 1.4155
Quantity 2.2 L
Physical Form Liquid
Grade Technical
Chemical Name or Material Nitric Acid, Technical 42 Degrees Baume’