missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid (Certified ACS Plus), Fisher Chemical™

Catalog No. A200S212
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Safe-Cote™ Glass
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A200S212 2.5 L Safe-Cote™ Glass
A200500 500 mL Glass Bottle
A200S500 500 mL Safe-Cote™ Glass
A200212 2.5 L Glass Bottle
A200C212 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A200S212 Supplier Thermo Fisher Scientific Supplier No. FLA200S212
Only null left

Nitric Acid material may darken during storage due to photochemical reaction

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH -1.0
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40g/cm³
Heavy Metals (as Pb) 0.2ppm max.
Identification Pass Test
Packaging Safe-Cote™ Glass
Residue after Ignition 2ppm max.
Vapor Pressure 0.94 kPa at 20°C
Normality about 15.8
Assay Percent Range 70%
Quantity 2.5 L
Physical Form Liquid
UN Number 2031
Formula Weight 63.01
Percent Purity 68.0 to 70.0% (w/w)
Grade Certified ACS Plus
Chemical Name or Material Nitric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:4
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive