missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, Certified, 6.0N ±0.1N, LabChem™

Catalog No. p-5388859
Change view
Click to view available options
Quantity:
500 mL
1 L
Catalog No. Quantity
LC178701 500 mL
LC178702 1 L
2 options

Catalog No. LC178701

Supplier: LabChem LC178701

Only null left

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Concentration 6.0N ±0.1N
CAS Max % 68, 32
Density 1.2g/mL
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.2g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Nitrogen oxides; Thermal decomposition generates: Corrosive vapors
Quantity 500 mL
Physical Form Liquid
Linear Formula HNO3
UN Number UN2031
Solubility Information Soluble in water
Formula Weight 63.01
Grade Certified
Chemical Name or Material Nitric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature