missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, Certified, 10.0% (v/v) ±0.5%, LabChem™

Catalog No. LC177301
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC177301 500 mL
LC177302 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC177301 Supplier LabChem Supplier No. LC177301
Only null left

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
CAS Max % 90.5, 9.5
Density 1.05g/mL
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.05g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Nitrogen oxides; Thermal decomposition generates: Corrosive vapors
Quantity 500 mL
Physical Form Liquid
Linear Formula HNO3
UN Number UN2031
Solubility Information Soluble in water
Formula Weight 63.01
Percent Purity 10.0% ±0.5% (v/v)
Grade Certified
Chemical Name or Material Nitric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature